Learn More
International Enviroguard™ GammaGuard CE™ Sterile Coveralls
Catalog No. 19472810 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-810 Large
19-472-812 Small
19-472-811 Medium
19-472-813 X-Large
19-472-807 2X-Large
19-472-808 3X-Large
19-472-809 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-472-810 Supplier International Enviroguard™ Supplier No. CE11019CISL
Add to Cart
Add to Cart
CE sterile coveralls offer enhanced safety with attached hood and boots along with a storm flap over the zipper.
  • Made of a comfortable fabric that protects against various liquids and particulates
  • Individually packaged and sterilized to a SAL of 10-6
  • Low-linting fabric helps reduce the release of unwanted particulates in the workspace
  • Coveralls are designed with tunnelized elastic that contains particle shed
  • Serged seams eliminate raw edges that can create and release additional particulate contamination
  • Attached hood and boots encourages quicker and easier donning processing
  • Elastic wrists and ankles help prevent particulates from entering or exiting the suit
  • Storm flap over the zipper adds an extra layer of protection
  • Convenient thumb loops are included to keep sleeves from riding up
  • Ankle ties secure the attached boots
  • Sterile-Don™ packaging protects against contamination during donning
  • GammaGuard CE™ helps keep the wearer cooler
  • Meets high manufacturing standards - made in an ISO-certified facility
  • Individually Packaged
Specifications
Product Type Cleanroom Coveralls
Size Large
Gender Unisex
Material Proprietary Laminate Non-woven Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic, Attached Boots
Cuff Style Elastic, Thumb Loop
Neck Style Hooded
Hood Type Attached, Tunnelized Elastic Face
Closure Type Zipper with Storm Flap
Seam Type Serged
Sterility Sterile
Protection Class General
Resistance Abrasion, Caustics, Liquid, Particle, Water
Back Style Tunnelized Elastic Back
Model CE11019CIS
Quantity 25/Cs.
Style Coveralls