Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell GAMMEX™ PI Hybrid Micro - Nonlatex Surgical Gloves

Catalog No. 17720509 Shop All Ansell Products
Change view
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-720-509 9
17-720-502 5.5
17-720-503 6
17-720-504 6.5
17-720-505 7
17-720-506 7.5
17-720-507 8
17-720-508 8.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-720-509 Supplier Ansell Supplier No. 340002090
  • $1,421.00
    $911.65 / Case of 200

    Save $509.35

    36% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Surgical Gloves that is suitable for delicate procedures requiring high tactile sensitivity and dexterity, protects from latex Type I allergy and minimize chemical Type IV allergies in HCW's or patients.

  • Smooth with micro-textured finish: Delivers secure grip and handling of instruments
  • Brown color glove: Suitable as an underglove
  • HYBRID Technology: Delivers latex-like comfort and high durability
  • Enhanced comfort and durability: Combining polyisoprene and neoprene to deliver a latex-like comfort and heightened durability
  • Heightened sensitivity: Being up to 24% thinner than Ansell's standard nonlatex surgical gloves, GAMMEX™ PI Hybrid Micro offer exceptional level of tactile sensitivity and dexterity for delicate procedures that requires utmost precision
  • Double-gloving versatility: This brown micro glove is suitable to be donned either as an outer glove; or inner glove when double gloving for easy breach detection
  • Reduced allergy risks: This synthetic blend material eliminates Type I latex allergy risks, and is free from Zinc 2-mercaptobenzothiazole-free (ZMBTfree), diphenylguanidinefree (DPG-free) and cetylpyridinium chloride-free (CPC-free) to minimize the risk of chemical Type IV allergies and sensitivities to these chemical accelerators
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM D3577, EN ISO 374-5:2016, EN ISO 21420:2020
Certifications/Compliance (Europe) EN 16523-1, EN 455 1-4, EN 420:2003 + A1:2009
Accelerator Free Yes
Applications Healthcare
AQL 0.65
Color Brown
Disposable Single-use
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Polyisoprene, Neoprene
Powdered Powder Free
Product Type Surgical Gloves
Size 9
Sterility Sterile
Manufacturing/QMS Audit Standards EN 556, ISO 11137-Part 1, ISO 13485, ISO 14001, ISO 9001
Packaging Type Polybag/Cardboard Box
Quantity 50 Pairs/Box
Tensile Strength Before aging ≥ 17
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.