Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell GAMMEX™ PI Plus Glove-in-Glove System

Catalog No. 17103775 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
7.5
8
8.5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-103-775 7
17-103-773 6
17-103-776 7.5
17-103-777 8
17-103-778 8.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-103-775 Supplier Ansell Supplier No. 340082070
Only null left
Add to Cart
Add to Cart

Synthetic, pre-donned double gloving system of two polyisoprene (PI) gloves in one, making it easy to double glove in one don

  • Pre-Donned Double-Gloving System: Saves valuable donning time
  • 50% Less Inner Packaging: Reduced environmental footprint
  • Skin-Friendly PI Formulation: No sensitization to known allergens*
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AS/NZS 4179|ASTM D3577|ASTM D7160|ASTM F739|EN 16523-1|EN 455 1-4|EN ISO 21420:2020|EN ISO 374-1:2016|EN ISO 374-5:2016|ISO 10282|JIS T9107
Accelerator Free Yes
Applications Laboratory
AQL 0.65
Color Green
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 16.93 ± 1.97 mil
Finger Thickness (Metric) 0.43 ± 0.05 mm
Hand Specific
Latex Free No
Material Polyisoprene
Palm Thickness (English) 15.55 ± 1.97 mil
Palm Thickness (Metric) 0.40 ± 0.05 mm
Powdered Powder Free
Size 7
Sterility Sterile
Thickness (English) 15.55 ± 1.97 mil
Thickness (Metric) 0.40 ± 0.05 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.