Learn More
Medchemexpress LLC HY-13637 100mg , Ganciclovir CAS:82410-32-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13637100MG
Encompass_Preferred
HY-13637 100mg Ganciclovir CAS: 82410-32-0 Ganciclovir (BW 759), a nucleoside analogue, is an orally active antiviral agent with activity against CMV. Ganciclovir also has activity in vitro against members of the herpes group and some other DNA viruses. Ganciclovir inhibits the in vitro replication of human herpes viruses (HSV 1 and 2, CMV) and adenovirus serotypes 1, 2, 4, 6, 8, 10, 19, 22 and 28. Ganciclovir has an IC50 of 5.2 μM for feline herpesvirus type-1 (FHV-1). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0084