Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ganciclovir Hydrate 98.0+%, TCI America™
SDP

Catalog No. G031525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G031525G 25 g
G03155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G031525G Supplier TCI America Supplier No. G031525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82410-32-0
Molecular Formula C9H13N5O4
Molecular Weight (g/mol) 255.234
MDL Number MFCD00870588
InChI Key IRSCQMHQWWYFCW-UHFFFAOYSA-N
Synonym ganciclovir, gancyclovir, cytovene, vitrasert, hydroxyacyclovir, ganciclovirum, zirgan, ganciclovir sodium, cymevene, hhemg
PubChem CID 3454
ChEBI CHEBI:465284
IUPAC Name 2-amino-9-(1,3-dihydroxypropan-2-yloxymethyl)-3H-purin-6-one
SMILES C1=NC2=C(N1COC(CO)CO)NC(=NC2=O)N

Specifications

Melting Point 250°C
Color White
Quantity 25 g
Formula Weight 255.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ganciclovir Hydrate
RTECSNumber MF8407000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.