Learn More
Apexbio Technology LLC GANT61, 5mg. Cas: 500579-04-4 MFCD: MFCD14635408. GLI antagonist
SDP
Supplier:  Apexbio Technology LLC A16155
Encompass_Preferred
GANT61 is a selective inhibitor of GLI with IC50 value of 13.56 M (NBL-W-S cells, 48 hours) [1].GLI genes, GLI1 and GLI2, are transcription activator and regulate target genes at the distal end of the canonical Hedgehog (HH) signaling pathway (SHH->PTCH->SMO->GLI). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4700
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.