Learn More
Selleck Chemical LLC Gap19-5mg
SDP
Supplier:  Selleck Chemical LLC S69465MG
Encompass_Preferred
Gap19 is a nonapeptide derived from the cytoplasmic loop (CL) of Cx43 and acts as a selective connexin 43 (Cx43) hemichannel blocker. Gap19 blocks Cx43 hemichannels without inhibiting gap junctions (GJs) at IC50 of 6.5 μM when applied intracellulary.
Catalog No. 50-238-9933
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.