Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7204373 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7394 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7394 Supplier Proteintech Group Inc Supplier No. 117111AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Required for ribosome biogenesis and telomere maintece. NOLA1 is part of the H/ACA small nucleolar ribonucleoprotein (H/ACA snoRNP) complex, which catalyzes pseudouridylation of rRNA. This involves the isomerization of uridine such that the ribose is subsequently attached to C5, instead of the normal N1. Each rRNA can contain up to 100 pseudouridine ("psi") residues, which may serve to stabilize the conformation of rRNAs. NOLA1 may also be required for correct processing or intranuclear trafficking of TERC, the RNA component of the telomerase reverse transcriptase (TERT) holoenzyme.

Specifications

Antigen GAR1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene GAR1
Gene Accession No. Q6AYA1, Q9CY66, Q9NY12
Gene Alias GAR1, NOLA1, snoRNP protein GAR1
Gene Symbols GAR1
Host Species Rabbit
Immunogen GAR1 Fusion Protein Ag2282
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499709, 54433, 68147
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.