Learn More
Eisco™ Garage Physics Center of Mass Demonstration Kit
SCI_ED
Catalog No. S24325 Shop All Eisco Products
Change view
Click to view available options
Physics Center of Mass Demonstration Kit
Catalog No.
S24325 Physics Center of Mass Demonstration Kit
1 options
Catalog No. S24325 Supplier Eisco™ Supplier No. GP00003
Add to Cart
Add to Cart
Learning mathematical concept of Center of Mass, allowing user to calculate and move the center of mass.
  • DIY assembly and activity
  • Kit contains: Storage box in image, Slotted Mass Set, protractor, high strain rubber bands (3), engineering balancing boards (2) and instructions
Specifications
Product Type Kit, Center of Mass
For Use With (Application) Gain a conceptual then mathematical knowledge of center of mass
Includes Slotted mass set, protractor, high strain rubber bands (3), engineering balancing boards (2), and instructions