Learn More
Eisco™ Garage Physics: Projectile Motion Slingshot Kit
SCI_ED
Catalog No. S27625 Shop All Eisco Products
Change view
Click to view available options
For Use With (Application):
Studying Projectile Motion
Catalog No. For Use With (Application)
S27625 Studying Projectile Motion
1 options
Catalog No. S27625 Supplier Eisco™ Supplier No. GP00014
Add to Cart
Add to Cart
The Garage Physics: Projectile Motion Kit allows students to explore projectile motion with a rubber-band slingshot!
  • Allows students of all ages to explore and visualize Hooke's Law, velocity, kinetic energy, gravity, and projectile motion
  • Students can use the included ping pong ball, or other light weight projectiles of their own design
  • The included stand allows exploration of launch angles from 0° to 45° in increments of 5°
  • Launches are highly repeatable giving consistent experimental results
  • Introduces and demonstrates real world STEM concepts
Specifications
Product Type Kit, Projectile Motion
For Use With (Application) Studying Projectile Motion
Includes Slingshot Frame, Slingshot Frame Supports, Release Mechanism Support, Angled Stands, Angled Stand Support, Rubber Bands, Ping Pong Ball, Leather Pouch, Cotter Pin, Cotter Pin String, Cardboard Storage Box, Instructions for Assembly, and Experimental Activ