Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Garage Physics Pythagorean Theorem Puzzle Kit
SCI_ED

Catalog No. S24326 Shop All Eisco Products
Change view
Click to view available options
Pythagorean Theorem Puzzle Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S24326 Pythagorean Theorem Puzzle Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24326 Supplier Eisco™ Supplier No. GP00004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Activity guide covers from elementary school through middle school, high school up to Einsteins proof of Pythangorean Theorem.

  • DIY assembly and activity
  • Kit contains 7-piece puzzle and instruction guide
  • Young children use kit to explore basic shapes and similarity
  • Elementary school children solve theorem by forming squares representing squares of hyotenuse and shorter sides
  • Middle school students use puzzle to work out 3 algebraic proofs of theorem
  • High school students use to explore units of measure and reason out Einstein proof of theorem

Specifications

Product Type Kit, Pythagorean Theorem Puzzle
Grade K to 12
Includes Seven piece puzzle
Grade Level Elementary, Intermediate - Middle School, High School
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.