Learn More
Eisco™ Garage Physics: Soda Center of Mass Kit
SCI_ED
Catalog No. S27624 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
S27624 1 Kit
1 options
Catalog No. S27624 Supplier Eisco™ Supplier No. GP00008
Add to Cart
Add to Cart
The Garage Physics: Soda Center of Gravity Kit allows students to explore the key concepts of balance, center of mass, and stability
  • Explore physical concepts surrounding balance, stability, and center of mass
  • Introduces and demonstrates real world STEM concepts
  • Designed to be accessible to small children while having sufficient depth to engage high-school students
  • Allows students of all ages to explore and visualize center of mass in dramatic fashion
  • Demonstration involves balancing a full, two liter bottle cantilevered at an angle with only indirect support
Specifications
Product Type Kit, Soda Center of Mass
For Use With (Application) Teaching Key Concepts Such as Balance, Center of Mass, and Stability
Includes Balance Board, Lever Arm, Pencil, Cardboard Storage Box, Assembly Instructions, and Experimental Activities
Quantity 1 Kit