Learn More
Eisco™ Garage Physics Torque Demonstration Kit
SCI_ED
Catalog No. S24324 Shop All Eisco Products
Change view
Click to view available options
Torque Demonstration kit
Catalog No.
S24324 Torque Demonstration kit
1 options
Catalog No. S24324 Supplier Eisco™ Supplier No. GP00002
Add to Cart
Add to Cart
Learning concept of torque by moving a mass set down a rod while trying to balance the main road
  • DIY assembly and activity
  • Kit contains: Storage Box in image, 1000g mass set, custom washer, 2 dowels, 2 bolts, 2 nuts, steel ball and instructions
Specifications
Product Type Torque Demonstration
For Use With (Application) Learn about mechanical advantage and torque
Includes Storage box, 1000 g mass set, custom washer, 2 dowels, 2 bolts, 2 nuts, steel ball and instructions