Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gardner Color Standard #9, Ricca Chemical
SDP

Catalog No. 32260916 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
322609-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 322609-16 Supplier Ricca Chemical Company Supplier No. 3226.0916
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10025-77-1 , 7791-13-1 , 7647-01-0 , 7732-18-5
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Packaging Amber Glass Bottle
CAS Min % 89.0, 5.44, 3.19, 0.84
CAS Max % 92.0, 5.66, 3.32, 0.87
Physical Form Liquid
Color Yellow
pH Acidic
For Use With (Equipment) Water Analyzer
Solubility Miscible with water
Solution Type Calibration Solution
Type Color Reference Standard
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Gardner Color Standard #9
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.