Learn More
Alpco Ltd GeneProof Gardnerella vaginalis PCR Kit
SDP
Supplier:  Alpco Ltd 60GV002025
Encompass
The GeneProof Gardnerella vaginalis PCR Kit can be combined with other tests within the STI panel to provide a comprehensive investigation of STIs. 25 tests. For Research Use Only in North America.
Catalog No. NC2885440