Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Garivulimab Recombinant Monoclonal Antibody

Catalog No. MA559110
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA559110 1 mg
MA559109 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA559110 Supplier Invitrogen™ Supplier No. MA559110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Monoclonal Antibody

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Garivulimab, also known as BGB-A333, is a humanized IgG1 monoclonal antibody that targets programmed death-ligand 1 (PD-L1, also known as CD274). CD274 is a gene encoding a protein critical for immune checkpoint regulation. Located on chromosome 9, this protein is expressed on the surface of various cell types, including tumor cells, immune cells, and normal tissues. PD-L1 interacts with the PD-1 receptor on T cells to modulate immune responses, particularly by promoting self-tolerance and preventing autoimmunity. In the context of cancer, aberrant expression of CD274 allows tumor cells to evade immune surveillance, leading to immune escape. This mechanism has made CD274 a significant target for cancer immunotherapies, particularly immune checkpoint inhibitors such as pembrolizumab and nivolumab, which block the PD-1/PD-L1 interaction to restore anti-tumor immunity. Research on CD274 continues to reveal its role in various malignancies and its potential as a prognostic and therapeutic biomarker.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Garivulimab Humanized
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Host Species Human
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.