Learn More
Thermo Scientific™ Gas Chromatography 3-Port SilFlow Device
Catalog No. 60201398 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1/Ea.
Catalog No. Quantity
60-201-398 1/Ea.
1 options
Catalog No. 60-201-398 Supplier Thermo Scientific™ Supplier No. 60201398
Add to Cart
Add to Cart
Switch your GC without the need to vent using the Thermo Scientific™ 3-Port SilFlow™ Device.
Switch your GC without the need to vent using the Thermo Scientific™ 3-Port SilFlow™ Device. This device comprises a gas module with microfluidics and Thermo Scientific™ SilTite™ FingerTite fittings for easy set up and a reliable seal.
TRUSTED_SUSTAINABILITY
Specifications
Type 3-Port SilFlow MCD
Description 3-Port SilFlow MCD (0.25/0.32)
Quantity 1/Ea.
For Use With (Equipment) Gas Chromatography Systems
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.