Learn More
Thermo Scientific™ Gas Chromatography 3-Port SilFlow Replacement Parts
Catalog No. 29063466
Change view
Click to view available options
Type:
Ferrule
FingerTite Tool
Silflow Nuts
For Use With (Equipment):
3-Port SilFlow Device
OD 0.36 mm (ID 0.25mm) Columns
OD 0.45 mm (ID 0.32mm) Columns
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
29-063-466 Ferrule OD 0.36 mm (ID 0.25mm) Columns
290SF302 Silflow Nuts 3-Port SilFlow Device
29-063-467 Ferrule OD 0.45 mm (ID 0.32mm) Columns
60-201-401 FingerTite Tool 3-Port SilFlow Device
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 29-063-466 Supplier Thermo Scientific™ Supplier No. 29063466
Add to Cart
Add to Cart
Switch your GC column in GC-MS without having to vent.
Maintain reliability with 3-Port SilFlow replacement parts when you place your next order.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule
Description SilFlow fingertight ferrules
Quantity 10 Pack
For Use With (Equipment) OD 0.36 mm (ID 0.25mm) Columns
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.