Learn More
Thermo Scientific™ Gas Chromatography 3-Port SilFlow Replacement Parts
Shop All Thermo Scientific ProductsDescription
Switch your GC column in GC-MS without having to vent.
Maintain reliability with 3-Port SilFlow replacement parts when you place your next order.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
| Type | Ferrule |
| Description | SilFlow fingertight ferrules |
| For Use With (Equipment) | OD 0.45 mm (ID 0.32mm) Columns |
| Quantity | 10 Pack |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.