Learn More
Thermo Scientific™ Gas Chromatography 3-Port SilFlow Replacement Parts
Catalog No. 60201401 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Ferrule
FingerTite Tool
Silflow Nuts
For Use With (Equipment):
3-Port SilFlow Device
OD 0.36 mm (ID 0.25mm) Columns
OD 0.45 mm (ID 0.32mm) Columns
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
60-201-401 FingerTite Tool 3-Port SilFlow Device
29-063-466 Ferrule OD 0.36 mm (ID 0.25mm) Columns
290SF302 Silflow Nuts 3-Port SilFlow Device
29-063-467 Ferrule OD 0.45 mm (ID 0.32mm) Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-201-401 Supplier Thermo Scientific™ Supplier No. 60201401
Add to Cart
Add to Cart
Switch your GC column in GC-MS without having to vent.
Maintain reliability with 3-Port SilFlow replacement parts when you place your next order.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type FingerTite Tool
Quantity 1 Each
For Use With (Equipment) 3-Port SilFlow Device
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.