Learn More
Description
- Meets USGS high-hazard seismic zone requirements when properly mounted
- Recommended for indoor or outdoor bulk cylinder storage
- Surround cylinders with 2-in (51-mm) square steel structural tubing
- Continuously seam welded at all junction points
- Sealed with exterior grade powder-coat paint for long lasting strength, durability and dependability
- Tops ground smooth and capped with high-density polyethylene (HDPE) caps to prevent corrosion and injuries
- Features two levels of welded steel chains for each cylinder and clasps to connect the chains to the rack, ensuring a snug fit
- Pre-drilled 0.531-in (14-mm) diameter floor mounting holes included on lower tube structure
- For seismic installations, add Model 35350 S4-rated anchor bolts (not included)
Compliance
OSHA, CGA P1, NFPA55
Warranty
1 YEAR
Specifications
Specifications
| Holds | 2 Cylinders: 1 Wide by 2 Deep |
| Material | Steel |
| Color | Black |
| For Use With (Equipment) | Gas cylinder |
| Height (Metric) | 762 mm |
| Length (Metric) | 406 mm |
| Weight (English) | 37 lb. |
| Width (Metric) | 406 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.