Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Gas Cylinder Barricade Rack

Catalog No. 19085780 Shop All Justrite Safety Group Products
Change view
Click to view available options
Holds:
1 Cylinder: 1 Wide by 1 Deep
10 Cylinders: 5 Wide by 2 Deep
12 Cylinders: 3 Wide by 4 Deep
12 Cylinders: 4 Wide by 3 Deep
15 Cylinders: 5 Wide by 3 Deep
16 Cylinders: 4 Wide by 4 Deep
2 Cylinders: 1 Wide by 2 Deep
2 Cylinders: 2 Wide by 1 Deep
20 Cylinders: 5 Wide by 4 Deep
3 Cylinders: 1 Wide by 3 Deep
3 Cylinders: 3 Wide by 1 Deep
4 Cylinders: 1 Wide by 4 Deep
4 Cylinders: 2 Wide by 2 Deep
4 Cylinders: 4 Wide by 1 Deep
5 Cylinders: 5 Wide by 1 Deep
6 Cylinders: 2 Wide by 3 Deep
6 Cylinders: 3 Wide by 2 Deep
8 Cylinders: 2 Wide by 4 Deep
8 Cylinders: 4 Wide by 2 Deep
9 Cylinders: 3 Wide by 3 Deep
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
19-085-780 2 Cylinders: 2 Wide by 1 Deep
19-085-776 1 Cylinder: 1 Wide by 1 Deep
19-085-777 2 Cylinders: 1 Wide by 2 Deep
19-085-778 3 Cylinders: 1 Wide by 3 Deep
19-085-779 4 Cylinders: 1 Wide by 4 Deep
19-085-781 4 Cylinders: 2 Wide by 2 Deep
19-085-782 6 Cylinders: 2 Wide by 3 Deep
19-085-783 8 Cylinders: 2 Wide by 4 Deep
19-085-784 3 Cylinders: 3 Wide by 1 Deep
19-085-785 6 Cylinders: 3 Wide by 2 Deep
19-085-787 9 Cylinders: 3 Wide by 3 Deep
19-085-789 12 Cylinders: 3 Wide by 4 Deep
19-085-790 4 Cylinders: 4 Wide by 1 Deep
19-085-791 8 Cylinders: 4 Wide by 2 Deep
19-085-792 12 Cylinders: 4 Wide by 3 Deep
19-085-793 16 Cylinders: 4 Wide by 4 Deep
19-085-794 5 Cylinders: 5 Wide by 1 Deep
19-085-795 10 Cylinders: 5 Wide by 2 Deep
19-085-796 15 Cylinders: 5 Wide by 3 Deep
19-085-797 20 Cylinders: 5 Wide by 4 Deep
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 19-085-780 Supplier Justrite™ Supplier No. 35100
Add to Cart
Add to Cart

Justrite™ Gas Cylinder Barricade Rack secures cylinders up to 12-in (305-mm) in diameter

  • Meets USGS high-hazard seismic zone requirements when properly mounted
  • Recommended for indoor or outdoor bulk cylinder storage
  • Surround cylinders with 2-in (51-mm) square steel structural tubing
  • Continuously seam welded at all junction points
  • Sealed with exterior grade powder-coat paint for long lasting strength, durability and dependability
  • Tops ground smooth and capped with high-density polyethylene (HDPE) caps to prevent corrosion and injuries
  • Features two levels of welded steel chains for each cylinder and clasps to connect the chains to the rack, ensuring a snug fit
  • Pre-drilled 0.531-in (14-mm) diameter floor mounting holes included on lower tube structure
  • For seismic installations, add Model 35350 S4-rated anchor bolts (not included)

Compliance

OSHA, CGA P1, NFPA55

Warranty and Services

1 YEAR

Specifications

Holds 2 Cylinders: 2 Wide by 1 Deep
Material Steel
Color Black
For Use With (Equipment) Gas cylinder
Height (Metric) 762 mm
Length (Metric) 762 mm
Weight (English) 41 lb.
Width (Metric) 762 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.