Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Gas Cylinder Barricade Rack for Cryogenic Cylinders

Catalog No. 19085798 Shop All Justrite Safety Group Products
Change view
Click to view available options
Holds:
1 Cylinder 22 in. dia.
1 Cylinder 33 in. dia.
2 Cylinders 22 in. dia.
2 Cylinders 33 in. dia.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
19-085-798 1 Cylinder 33 in. dia.
19-085-799 1 Cylinder 22 in. dia.
19-085-800 2 Cylinders 22 in. dia.
19-085-801 2 Cylinders 33 in. dia.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-085-798 Supplier Justrite™ Supplier No. 35184
Add to Cart
Add to Cart

Justrite™ Gas Cylinder Barricade Rack for Cryogenic Cylinders meets USGS high-hazard seismic zone requirements when properly mounted

  • Recommended for indoor or outdoor cryogenic cylinder storage
  • Surround cylinders with 2-in (51-mm) square steel structural tubing
  • Continuously seam welded at all junction points
  • Sealed with exterior grade powder-coat paint for long lasting strength, durability and dependability
  • Tops ground smooth and capped with high-density polyethylene (HDPE) caps to prevent corrosion and injuries
  • Features two levels of welded steel chains for each cylinder and clasps to connect the chains to the rack, ensuring a snug fit
  • Pre-drilled 0.531-in (14-mm) diameter floor mounting holes included on lower tube structure
  • For seismic installations, add Model 35350 S4-rated anchor bolts (not included)

Compliance

OSHA, CGA P1, NFPA55

Warranty and Services

1 YEAR

Specifications

Holds 1 Cylinder 33 in. dia.
Material Steel
Color Black
Height (Metric) 1016 mm
Length (Metric) 965 mm
Weight (English) 109 lb.
Width (Metric) 965 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.