Learn More
Justrite™ Gas Cylinder Forklift Pallet, End Loaded
Catalog No. 19085802 Shop All Justrite Safety Group Products
Change view
Click to view available options
Holds:
14 Cylinders
21 Cylinders
6 Cylinders
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
19-085-802 6 Cylinders
19-085-804 14 Cylinders
19-085-806 21 Cylinders
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-085-802 Supplier Justrite™ Supplier No. 35210
Add to Cart
Add to Cart
Justrite™ Gas Cylinder Forklift Pallet is end-load oriented to ensure secure forklift transport of multiple gas cylinders
  • Fully enclosed 7.75-in wide (197-mm) forklift channels with two way access
  • Textured finish on the pallet provides a non-skid surface
  • Secure a group of gas cylinders using the 10,000-lb (4,536-kg) DOT compliant strap that attaches to frame at either 12-, 24- or 36-in (305-, 610- or 915-mm) from the deck
  • Fully welded construction from 11-gauge (3-mm) hot-rolled steel and structural tube steel, powder-coated to ensure longevity and chemical resistance
Compliance
OSHA, CGA P1, NFPA55
Warranty and Services
1 YEAR
Specifications
Holds 6 Cylinders
Material Steel
Color Black
Description End loaded
Dimensions (L x W x H) 49.5 x30 x32 in.
Height (Metric) 965 mm
Weight (English) 62 lb.
Width (Metric) 610 mm