Learn More
Trajan Scientific and Medical Gas Filter Connecting Unit
SDP
The gas filter connecting unit has inlet and outlet connectors for the gas line and the system can be wall-mounted or fixed to a bench. Connecting units come with a 1, 2 or 4 filter capacity and are available for 1/4 in. or 1/8 in. gas lines
Supplier:  Trajan Scientific and Medical 1035044
 View more versions of this product
Catalog No. SG1035044
Description
Description
  • Easily installed into the gas line
  • Can be either wall-mounted or fixed to a bench
  • Available with a 1, 2 or 4 filter capacity
Specifications
Specifications
1/4 in. fittings
Gas filters
1/4 in.
4-position Connecting Unit
Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For research use only