Learn More
Cytiva Spare Parts and Accessories for 47 mm pressure filtration funnel - PTFE gasket (1/pkg)
Catalog No. 502063027 Shop All Cytiva Products
Encompass
High Quality, Corrosion-Resistant Funnel for Rapid Batch Filtration-Stainless steel construction offers excellent chemical resistance. -Convenient hand tightening makes filter changing easy. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3027 Supplier Cytiva Supplier No. 4284
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
High Quality, Corrosion-Resistant Funnel for Rapid Batch Filtration-Stainless steel construction offers excellent chemical resistance. -Convenient hand tightening makes filter changing easy. This product is not approved for use in medical, clinical, surgical or other patient applications.