Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD GasPak™ Supplies

Catalog No. L60630 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Type:
CO2 Indicator Strip
Dry Anaerobic Indicator Strip
Tube Holder
Quantity:
1 Each
100 Pk.
50 Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
L60630 Tube Holder 1 Each
B4371055 CO2 Indicator Strip 50 Pk.
B71051 Dry Anaerobic Indicator Strip 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L60630 Supplier BD Supplier No. 260630
Add to Cart
Add to Cart

Used with the GasPak™ EZ Gas Generating Systems. BD™ GasPak™ Supplies are used with these complete, integrated systems for rapid, waterless and catalyst-free gas generation. Can be used to support an anaerobic, microaerophilic or CO2 enriched environment.

GasPak™ EZ Gas Generating Systems are complete, integrated systems that rapidly produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.

Accessories include:

  • CO2 Indicator Strips
  • Dry Anaerobic Indicator Strips
  • Tube Holder
  • Catalyst Replacement Charges
  • Catalyst Reaction Chamber
TRUSTED_SUSTAINABILITY

Specifications

Type Tube Holder
Content And Storage 15°C to 30°C
Description Clear, plastic tube holder
For Use With (Application) Produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria
For Use With (Equipment) BD GasPak™
Holds 16.5 x 128 Tubes (13)
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.