Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ Gastrin Double Antibody RIA Kit

Catalog No. MP06B255017 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 x 100 Tubes
1 x 200 Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP06B255017 1 x 100 Tubes
MP06B255025 1 x 200 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP06B255017 Supplier MP Biomedical™ Supplier No. 06B255017

This product is regulated by the federal government, and may not be ordered online.

Quantitative Liquid Phase Radioimmunoassay

The Gastrin Double Antibody RIA Kit is for the quantitative determination of immunoreactive gastrins in serum.
TRUSTED_SUSTAINABILITY

Specifications

Description For the quantitative determination of immunoreactive gastrins in serum.
Product Type Kit and Assay
Sample Type Serum
Target Species Human
For Use With (Application) Endocrinology
Quantity 1 x 100 Tubes
Regulatory Status FDA Cleared (K951329S2)
Sample Volume 200 μL
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.