Learn More
Apexbio Technology LLC GASTRIN I HUMAN 1MG
SDP
Supplier:  Apexbio Technology LLC B53581
Encompass_Preferred
Gastrin I (human) (CAS 10047-33-3) is an endogenous regulatory peptide that stimulates gastric acid secretion through specific receptor interactions on gastric parietal cells By engaging these target receptors Gastrin I activates intracellular signaling cascades influencing proton pump activity and subsequent acid release In vitro Gastrin I serves as an experimental tool to investigate gastric acid secretion pathways receptor-mediated signal transduction and gastrointestinal physiology It is particularly relevant in research exploring gastrointestinal disorders and therapeutic intervention mechanisms
Catalog No. 50-224-0049
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.