Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gastrin Mouse anti-Human, Clone: 7G9C3, Proteintech
SDP

Catalog No. p-7213288 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6433 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6433 Supplier Proteintech Group Inc Supplier No. 603461IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Gastrin is a 17 amino acid peptide hormone and neurotransmitter widely distributed throughout the gastrointestinal (GI) tract and the central nervous system (CNS). Study of gastrin may be used to study the differential tissue expression and intracellular and subcellular localization of gastrin in neuroendocrine cells of the gastrointestinal tract, and in the CNS.

Specifications

Antigen Gastrin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7G9C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GAST
Gene Accession No. P01350
Gene Alias G14 Gastrin 6, G17 Gastrin 14, G34 Gastrin, G52 Big gastrin, G6, GAS, GAST, gastrin, Gastrin 17, Gastrin 34, Gastrin component I Gastrin 52, Gastrin component II, Gastrin component III
Gene Symbols GAST
Host Species Mouse
Immunogen Gastrin Fusion Protein Ag12795
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2520
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.