Learn More
Enzo Life Sciences Gastrin polyclonal antibody (25µl)
SDP
Supplier:  Enzo Life Sciences BMLGA11670025
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 18-34 of gastrin I (gastrin-17; gastrin-34). Source: Whole rabbit serum. Species reactivity: Human, Rat Porcine. Applications: IHC. Formulation: Liquid. Neat serum.
Catalog No. 50-201-0893
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.