Learn More
Eurofins Discoverx Gastrin Releasing Peptide, 0.5 mg. (92-1057)
Supplier:  Eurofins Discoverx 921057
Encompass_Preferred
Gastrin-releasing peptide (GRP) is a 27 amino acid regulatory neuropeptide that is implicated in both physiological and pathophysiological processes. It is involved in inducing the release of gastrin from G cells of the stomach as well as the regulation of gastric acid secretion and enteric motor function. It also has a role in the circadian system, relaying light signals to the master circadian oscillator in the hypothalamus. Negative feedback regulator of fear, and may also mediate stress. Acts as an agonist for GRPR. Provided as 0.5 mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9598
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.