Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gastrodin 98.0+%, TCI America™
SDP

Catalog No. G04681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04681G 1 g
G0468200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G04681G Supplier TCI America Supplier No. G04681G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62499-27-8
Molecular Formula C13H18O7
Molecular Weight (g/mol) 286.28
MDL Number MFCD00272169
InChI Key PUQSUZTXKPLAPR-UJPOAAIJSA-N
Synonym 4-(Hydroxymethyl)phenyl beta-D-Glucopyranoside
PubChem CID 115067
ChEBI CHEBI:80828
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[4-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
SMILES C1=CC(=CC=C1CO)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 155°C
Color White
Quantity 1 g
Formula Weight 286.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Gastrodin
RTECSNumber LZ5776885
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.