Learn More
Therapak™ Gastroenterology (GI) Biopsy Collection and Transport Kits
Catalog No. 22131462 Shop All Therapak Corporation Products
Change view
Click to view available options
Product Type:
6-Vial GI Biopsy Transport Kit
Catalog No. Product Type
22-131-462 6-Vial GI Biopsy Transport Kit
1 options
This item is not returnable. View return policy
Catalog No. 22-131-462 Supplier Therapak™ Supplier No. 38874
This item is not returnable. View return policy
Features standardized components to maximize the ease of collecting and packing specimens for transport
Primary receptacles in the form of 20mL jars are 95kPa certified for transport in compliance with DOT and IATA standards for leak resistant transportation. A set of jar labels with the location ID is included in the kit to ensure proper placement of the specimen jars at the point of packing and identification once received at the test location. Customization of these kits is available upon request.
  • Comes with either two or six 20mL jars pre-filled with 10mL of formalin solution
  • Jars are secured by a corrugated insert and placed into the provided zip-style biohazard bag with absorbent material for transport
  • Corrugated inserts are printed with the GI biopsy location ID for easy recognition
Specifications
Product Type 6-Vial GI Biopsy Transport Kit
No. per Case 20/Cs.