Learn More
Description
PA1-102 has been successfully used in Western blot, FACS, immunohistochemistry (paraffin) and immunofluorescence applications with human and mouse samples. PA1-102 can be used for immunofluorescence analysis of GATA4 in the endoderm derived from human iPSCs.
Specifications
Specifications
| Antigen | GATA4 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol, 1mg/mL BSA and 0.05% sodium azide |
| Gene | GATA4 |
| Gene Accession No. | P43694, Q08369 |
| Gene Alias | ASD2; DNA-binding protein GATA-GT2; GAT4; GATA binding protein 4; GATA4; Gata-4; GATA-binding factor 4; GATA-binding protein 4; MGC126629; TACHD; TOF; Transcription factor GATA-4; VSD1 |
| Gene Symbols | GATA4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.