Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GATAD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568586
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568586 Supplier Invitrogen™ Supplier No. PA568586
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.

Component of some chromatin complex recruited to chromatin sites methylated 'Lys-4' of histone H3 (H3K4me).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATAD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GATAD1
Gene Accession No. Q8WUU5
Gene Alias 2310031E19Rik; 2810047M21Rik; 8430439A17Rik; 9130430G15Rik; B330017N08Rik; CMD2B; ENSMUSG00000073241; GATA zinc finger domain containing 1; GATA zinc finger domain-containing 1-like protein; GATA zinc finger domain-containing protein 1; GATA zinc finger domain-containing protein 1-like protein; Gatad1; im:7150682; ocular development associated; ocular development-associated gene protein; ODAG; RG083M05.2; RG83M5.2; si:dkey-231l1.7; Unknown (protein for MGC:136582); zgc:136582
Gene Symbols GATAD1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human GATAD1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57798
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.