Learn More
Invitrogen™ GATAD2A Polyclonal Antibody

Description
Suggested positive control: HeLa or RAW.
Specifications
Specifications
| Antigen | GATAD2A |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with 0.05% sodium azide |
| Gene | GATAD2A |
| Gene Accession No. | Q86YP4, Q8CHY6 |
| Gene Alias | 1110066C11Rik; BC031407; C80248; GATA zinc finger domain containing 2A; GATA zinc finger domain-containing protein 2A; GATAD2A; Hp66alpha; MGC29315; MGC36975; p66 alpha; p66a; p66alpha; RGD1310596; transcriptional repressor p66 alpha; Transcriptional repressor p66-alpha |
| Gene Symbols | GATAD2A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.