Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GATAD2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675670
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-670 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-670 Supplier Invitrogen™ Supplier No. PA141333
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: HeLa or RAW.

This gene encodes a zinc finger protein transcriptional repressor. The encoded protein is part of the methyl-CpG-binding protein-1 complex, which represses gene expression by deacetylating methylated nucleosomes. Mutations in this gene are linked to intellectual disability and dysmorphic features associated with cognitive disability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATAD2B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene GATAD2B
Gene Accession No. Q8VHR5, Q8WXI9
Gene Alias AL118180; C430014D17Rik; GATA zinc finger domain containing 2B; GATA zinc finger domain-containing protein 2B; GATAD2B; KIAA1150; mKIAA1150; MRD18; p66/p68; P66beta; p68; RGD1308533; transcription repressor p66 beta component of the MeCP1 complex; transcription repressor p66 component of the MeCP1 complex; transcriptional repressor p66-beta
Gene Symbols GATAD2B
Host Species Rabbit
Immunogen Synthetic peptide corresponding to aa 445-460 of mouse p66b.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229542, 57459
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.