Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Gate Valve Lockout 2.5 - 5 in Wheel Dia.

Catalog No. 19104078 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-104-078 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-104-078 Supplier Brady™ Supplier No. 73715

Cover the valve handle with the gate valve lockout device and apply a personal padlock and lockout tag to ensure employee safety

Wheel Dia.: 2.5 - 5 in

Specifications

Product Type Hinged Lockout Device
Color Red
Material Polypropylene
Certifications/Compliance OSHA 1910.147
Diameter (English) 2.78 in.
Quantity 1 Ea.
Width (Metric) 140 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.