Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gatifloxacin, G-Biosciences
SCI_ED

Catalog No. S139843
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139843 1 g
S157226 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139843 Supplier G-Biosciences Supplier No. RC1032
Add to Cart
Add to Cart

Gatifloxacin is a fourth generation fluoroquinolone antibiotic with a significant degree of activity towards gram-negative bacteria and enhanced activity toward gram-positive bacteria.

Gatifloxacin is a fourth generation fluoroquinolone antibiotic with a significant degree of activity towards gram-negative bacteria and enhanced activity toward gram-positive bacteria. It is also potent in vivo against Staphylococcus aureus, Clostridium and many other anaerobic bacteria. Gatifloxacin has also exhibited strong bactericidal efficiency in penicillin resistant and sensitive Streptococcus pneumoniae and S. aureus.

Fluoroquinolones are a group of broad-spectrum antibiotics that inhibit DNA synthesis. These antibiotics prevent unwinding of double-stranded DNA by binding DNA gyrase or DNA topoisomerase.

Chemical Identifiers

Molecular Weight (g/mol) 401.41 g/mol

Specifications

Boiling Point 607.8°C
Quantity 1 g
Percent Purity >98%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.