Learn More
Med Vet International Sponge Gauze, 3"X3", 200/Bag, 12Ply
Supplier:  Med Vet International JOR193B
Encompass_Preferred
Our sponge gauze is 100 percent pure cotton and comes in a 20 x 12 mesh. It is fully bleached and meets the USP standard.
Catalog No. 50-283-0690
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.