Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBP1 Recombinant Rabbit Monoclonal Antibody (K01_2M50)
GREENER_CHOICE

Catalog No. MA564069
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA564069 100 μL
MA565528 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA564069 Supplier Invitrogen™ Supplier No. MA564069
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: LLKEGFQKES RIMK.

GBP1 or guanlyate binding protein 1 is a 68 kDa GTPase and member of the GTPase1 family of molecules. GBP1 expression is induced by Interferon g as well as other inflammatory cytokines including IL-1a, IL-1b and TNF-a. Induction of GBP1 expression by poly I:C, a prototypical TLR3 ligand, as well as GBP1 expression dependency on NF-kB and IFNa response elements implicate TLR pathway involvement. Inversely proportional expression of GBP1 and MMP-1 as well as other related molecules to adhesion and migration show a clear functional role of GBP1 in suppressing angiogenesis and migration especially of endothelial cells. As a regulatory molecule it is centrally involved in signal transduction of inflammatory and cell migration networks.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2M50
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene GBP1
Gene Accession No. P32455
Gene Alias GBP1; Gbp-1; Gbp2b; GTP-binding protein 1; guanine nucleotide-binding protein 1; guanylate binding protein 1; guanylate binding protein 1, interferon-inducible; guanylate binding protein 1, interferon-inducible, 67kDa; guanylate binding protein 2b; guanylate nucleotide binding protein 1; guanylate-binding protein 1; huGBP-1; interferon-gamma-inducible protein MAG-1; interferon-induced guanylate-binding protein 1; Mag-1; Mpa1; Mpa-1
Gene Symbols GBP1
Host Species Rabbit
Immunogen A synthetic peptide of human GBP1 (aa 560-573).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2633
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.