Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBP2 Monoclonal Antibody (OTI5C8)
GREENER_CHOICE

Catalog No. PIMA524980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524980 Supplier Invitrogen™ Supplier No. MA524980
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GBP2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

Binds GTP, GDP and GMP. Hydrolyzes GTP very efficiently; GDP rather than GMP is the major reaction product.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBP2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI5C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GBP2
Gene Accession No. P32456
Gene Alias GBP2; GBP-2; GTP-binding protein 2; guanine nucleotide-binding protein 2; guanylate binding protein 2; guanylate binding protein 2, interferon-inducible; guanylate nucleotide binding protein 2; Guanylate-binding protein 1; Guanylate-binding protein 2; interferon-induced guanylate-binding protein 2; isoprenylated 67 kDa protein; mGBP2; mGBP-2; p67
Gene Symbols GBP2
Host Species Mouse
Immunogen Full length human recombinant protein of GBP2 produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2634
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.