Learn More
Medchemexpress LLC HY-100968 5mg Medchemexpress, GBR 12783 (dihydrochloride) CAS:67469-75-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1009685MG
Encompass_Preferred
Medchemexpress, HY-100968 5mg GBR 12783 (dihydrochloride) CAS:67469-75-4 GBR 12783 dihydrochloride is a specific, potent and selective dopamine uptake inhibitor that inhibits the [3H]dopamine uptake by rat and mice striatal synaptosomes with IC50s of 1.8 nM and 1.2 nM, respectively. GBR 12783 dihydrochloride can improve memory performance and increase hippocampal acetylcholine release in rats. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0610
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.