Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GBR 12909 dihydrochloride
GREENER_CHOICESDP

Catalog No. 042150
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-215-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 04-215-0 Supplier Tocris Bioscience™ Supplier No. 0421/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective DA uptake inhibitor; also σ ligand

GBR 12909 dihydrochloride is a potent, competitive inhibitor of dopamine uptake (Ki = 1 nM for inhibition of striatal dopamine uptake). Has >100-fold lower affinity for the noradrenaline and 5-HT uptake carriers. Also a potent sigma ligand (IC50 = 48 nM). Centrally active following systemic administration.

Specifications

Chemical Name or Material 1-[2-[Bis-(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)piperazine dihydrochloride
CAS 67469-78-7
Assay Percent Range >98%
Quantity 50 mg
Solubility Soluble to 50mM in DMSO
Molecular Formula C28H32F2N2O.2HCl
Formula Weight Observed MW: 523.49

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.