Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ GBS Detect™
SDP

Catalog No. 23001736 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-736 10
23-001-790 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-001-736 Supplier Hardy Diagnostics™ Supplier No. A300
Add to Cart
Add to Cart
This item is not returnable. View return policy

GBS Detect™ is recommended for the isolation and differentiation by enhanced hemolysis of detection of gamma-hemolytic Group B Streptococcus from GBS enrichment broth procedures.

  • Optically clear polystyrene for distortion free viewing
  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding.
  • Packaged in sleeves of 10 plates in a breathable bag that prevents buildup of condensation and excess moisture

Specifications

Color Red
Content And Storage 2°C to 8°C
Additive Selective Agents, Hemolysis Inducing Agents
Detects Group B Streptococcus
Form Prepared Medium
Format Plate
Product Type Agar Plate
Sterility Sterile
Target Organism Class Gram-positive Cocci
Test Type Microbial Differentiation
Fill Volume 17 mL
Formulation Pancreatic Digest of Casein - 15.0 gm, Peptic Digest of Soybean Meal - 5.0 gm, Sodium Chloride - 5.0 gm, Nucleic Acid - 3.0 gm, Selective Agents - 15.3 gm, Hemolysis Inducing Agents - 20.0 ml, Sheep Blood - 50.0 ml, Agar - 15.0 gm
For Use With (Application) Isolation and differentiation by enhanced hemolysis of detection of gamma-hemolytic Group B Streptococcus from GBS enrichment broth procedures
Incubation Conditions 35 ±2°C, Aerobic
Incubation Time 18 to 24 hr.
Material Polystyrene
No. per Pack 10
Packaging Type Bag
Quantity 10 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Streptococcus species
Protein Source Casein, Soybean Meal, Sheep Blood
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.