Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577984 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577984 Supplier Invitrogen™ Supplier No. PA577984
  • $640.00
    $494.65 / Each of 1

    Save $145.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Positive Control: U87-MG, SK-N-SH, IMR32, IMR32 nuclear, Neuro 2A, C8D30, NIH-3T3, Raw264.7, C2C12, mouse fetal brain, rat brain Predicted Reactivity: Bovine (96%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Altered expression of GBX2, part of the homeobox-containing human family of DNA-binding transcription factors, is associated with therapy failure and death in patients with multiple types of cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBX2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene GBX2
Gene Accession No. P48031, P52951
Gene Alias D130058E05Rik; gastrulation and brain-specific homeobox protein 2; gastrulation brain homeobox 2; Gbx2; Gbx-2; Homeobox protein GBX-2; LOW QUALITY PROTEIN: homeobox protein GBX-2; MMoxA; Stimulated by retinoic acid gene 7 protein; Stra7
Gene Symbols GBX2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GBX2. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114500, 14472, 2637
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.