Learn More
Enzo Life Sciences Guanylyl cyclase alpha 1 beta 1 (soluble) (human), (recombinant) (His-tag) (enriched) (50µl)
Supplier: Enzo Life Sciences ALX201180R050
Alternative name: GTP pyrophosphate lyase (cycling), sGC alpha 1 / sGC beta 1. Concentration: ≥1.5mg/ml (total protein). Formulation: Liquid. Enriched fraction in 50mM TEA, pH 7.4 containing 1mM EDTA/EGTA, 5mM GSH and 30% glycerol. Source: Produced in Sf9 insect cells. Fused at the C-terminus to a His-tag. Produced in a baculovirus expression system. Long Term Storage: -80°C. Use/Stability: We do not recommend to store for longer periods after opening and thawing of the product. Handling: Avoid freeze/thaw cycles.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.