Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek Corporation GC COLUMN 30M 0.53MM ID 20UM
SDP

Catalog No. NC2588195 Shop All Restek Corporation Products
Encompass_Preferred

RestekRt-Q-BOND PLOT Column, 30 m, 0.53 mm ID, 20 um

Catalog No. NC2588195 Supplier Restek Corporation Supplier No. 19742
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RestekRt-Q-BOND PLOT Column, 30 m, 0.53 mm ID, 20 um

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.