Learn More
Krackeler Scientific GC Inlet Liners
Specifications
Specifications
| Description | Straight, Glass Wool |
| Volume (Metric) | 990 μL |
| Includes | Glass wool |
| Product Type | Split Liner |
| Diameter (Metric) Inner | 4 mm |